Learn More
(R)-1-(2-Methoxyphenyl)ethylamine, ChiPros™, 99%, ee 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2766906
Description
The derivatives of the compound, are used as a potential ligand in developing radioiodinated antagonists for imaging of 5-HT1A receptors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe derivatives of the compound, are used as a potential ligand in developing radioiodinated antagonists for imaging of 5-HT1A receptors.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
(R)-1-(2-Methoxyphenyl)ethylamine | |
118°C (17 mmHg) | |
C9H13NO | |
5 g | |
Air Sensitive | |
Soluble in dimethyl sulfoxide. | |
COC1=CC=CC=C1C(C)N | |
151.21 | |
151.21 |
68285-23-4 | |
ee ≥98% | |
MFCD00671657 | |
UN2735 | |
r-1-2-methoxyphenyl ethanamine, r-2-methoxy-alpha-methylbenzylamine, chipros r , produced by basf, r-1-2-methoxyphenyl ethylamine, 1r-1-2-methoxyphenyl ethan-1-amine, 1r-1-2-methoxyphenyl ethan-1-amine, a-methylbenzylamine, 1r-1-2-methoxyphenyl ethylamine, r-2-methoxy-alpha-methylbenzylamine, r-2-methoxy-alpha-methylbenzylamine, r-1-2-methoxyphenyl ethylamine, chipros, r-1-2-methoxyphenyl ethylamine, chipros | |
VENQOHAPVLVQKV-UHFFFAOYNA-N | |
(1R)-1-(2-methoxyphenyl)ethanamine | |
853083 | |
99% |
Chemical Identifiers
68285-23-4 | |
151.21 | |
VENQOHAPVLVQKV-UHFFFAOYNA-N | |
853083 | |
COC1=CC=CC=C1C(C)N |
C9H13NO | |
MFCD00671657 | |
r-1-2-methoxyphenyl ethanamine, r-2-methoxy-alpha-methylbenzylamine, chipros r , produced by basf, r-1-2-methoxyphenyl ethylamine, 1r-1-2-methoxyphenyl ethan-1-amine, 1r-1-2-methoxyphenyl ethan-1-amine, a-methylbenzylamine, 1r-1-2-methoxyphenyl ethylamine, r-2-methoxy-alpha-methylbenzylamine, r-2-methoxy-alpha-methylbenzylamine, r-1-2-methoxyphenyl ethylamine, chipros, r-1-2-methoxyphenyl ethylamine, chipros | |
(1R)-1-(2-methoxyphenyl)ethanamine |
Safety and Handling
GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H331-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Store under Argon
RUO – Research Use Only