Learn More
(R)-(+)-1-(2-Bromophenyl)ethanol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5600303
Description
It is the most important dyestuff intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is the most important dyestuff intermediate.
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications
(R)-(+)-1-(2-Bromophenyl)ethanol | |
54°C to 56°C | |
MFCD00216591 | |
r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethan-1-ol, pubchem5686, r-+-1-2-bromophenyl ethanol, r-+-2-bromo-, a-methylbenzyl alcohol, benzenemethanol,2-bromo-a-methyl-, ar, r-+-2-bromo-alpha-methylbenzyl alcohol, r-+-2-bromo-alpha-methylbenzyl alcoh | |
+54° (c=1 in Chloroform) | |
(1R)-1-(2-bromophenyl)ethanol | |
2734868 | |
98% |
76116-20-6 | |
C8H9BrO | |
1 g | |
DZLZSFZSPIUINR-ZCFIWIBFSA-N | |
CC(C1=CC=CC=C1Br)O | |
201.063 | |
201.06 |
Chemical Identifiers
76116-20-6 | |
201.063 | |
DZLZSFZSPIUINR-ZCFIWIBFSA-N | |
2734868 | |
CC(C1=CC=CC=C1Br)O |
C8H9BrO | |
MFCD00216591 | |
r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethan-1-ol, pubchem5686, r-+-1-2-bromophenyl ethanol, r-+-2-bromo-, a-methylbenzyl alcohol, benzenemethanol,2-bromo-a-methyl-, ar, r-+-2-bromo-alpha-methylbenzyl alcohol, r-+-2-bromo-alpha-methylbenzyl alcoh | |
(1R)-1-(2-bromophenyl)ethanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only