missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-1-(2-Bromophenyl)ethanol, 98%

Manufacturer:  Alfa Aesar™ H5600306

 View more versions of this product

Catalog No. AAH5600306



Description

Description

Applications
It is the most important dyestuff intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

(R)-(+)-1-(2-Bromophenyl)ethanol
54°C to 56°C
76116-20-6
MFCD00216591
DZLZSFZSPIUINR-ZCFIWIBFSA-N
(1R)-1-(2-bromophenyl)ethanol
2734868
98%
+54° (c=1 in Chloroform)
5g
C8H9BrO
1r-1-2-bromophenyl ethan-1-ol
CC(C1=CC=CC=C1Br)O
201.063
201.06

Chemical Identifiers

76116-20-6
201.063
DZLZSFZSPIUINR-ZCFIWIBFSA-N
2734868
CC(C1=CC=CC=C1Br)O
C8H9BrO
MFCD00216591
1r-1-2-bromophenyl ethan-1-ol
(1R)-1-(2-bromophenyl)ethanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents