Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-1,2-Bis(diphenylphosphino)propane, 98%

$100.20 - $329.99

Chemical Identifiers

Chemical Name or Material (R)-(+)-1,2-Bis(diphenylphosphino)propane
CAS 67884-32-6
Molecular Formula C27H26P2
Formula Weight 412.46
MDL Number MFCD00012309
Synonym (R)-PROPHOS
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2107801

  SDS
alfa aesar™
B2107801
0.25g Each for $100.20

AAB2107803

  SDS
alfa aesar™
B2107803
1g Each for $329.99
Description & Specifications

Specifications

Formula Weight 412.46
Chemical Name or Material (R)-(+)-1,2-Bis(diphenylphosphino)propane
Assay 98%
Molecular Formula C27H26P2
MDL Number MFCD00012309
CAS 67884-32-6
Synonym (R)-PROPHOS
Melting Point 71° to 73°C
Health Hazard 1 Warning
Quantity 0.25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.