Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-1,2,3,4-Tetrahydroisoquinoline-3-methanol, 97%

$90.50 - $90.50

Chemical Identifiers

Chemical Name or Material (R)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
CAS 62855-02-1
Molecular Formula C10H13NO
Formula Weight 163.22
MDL Number MFCD02684327
Beilstein 7290561
Synonym (R)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6540106

 
alfa aesar™
H6540106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6540103

 
alfa aesar™
H6540103
1g Each for $90.50
Description & Specifications

Specifications

Formula Weight 163.22
Physical Form Crystalline
Assay 97%
Molecular Formula C10H13NO
MDL Number MFCD02684327
CAS 62855-02-1
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material (R)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
Beilstein 7290561
Synonym (R)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline
Quantity 5g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.