missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine, TCI America™

Manufacturer:  TCI America T292625G

 View more versions of this product

Catalog No. T292625G



Specifications

Specifications

(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine
Yellow
23357-46-2
MFCD00671629
JRZGPXSSNPTNMA-SNVBAGLBSA-N
(1R)-1,2,3,4-tetrahydronaphthalen-1-amine
7058072
Clear Liquid at 20°C
70°C
25g
C10H13N
r-1,2,3,4-tetrahydronaphthalen-1-amine, r---1,2,3,4-tetrahydro-1-naphthylamine, 1r-1,2,3,4-tetrahydronaphthalen-1-amine, r-1,2,3,4-tetrahydro-1-naphthylamine, 1-naphthalenamine, 1,2,3,4-tetrahydro-, 1r, r-1-amino-1,2,3,4-tetrahydronaphthalene, r-1, 2, 3, 4-tetrahydro-1-naphthylamine, r-1-aminotetraline, tetralin-1alpha-amine, r-1-aminotetralin
C1CC(C2=CC=CC=C2C1)N
147.221
147.22
≥98.0% (GC,T)

Chemical Identifiers

23357-46-2
147.221
JRZGPXSSNPTNMA-SNVBAGLBSA-N
7058072
C1CC(C2=CC=CC=C2C1)N
C10H13N
MFCD00671629
r-1,2,3,4-tetrahydronaphthalen-1-amine, r---1,2,3,4-tetrahydro-1-naphthylamine, 1r-1,2,3,4-tetrahydronaphthalen-1-amine, r-1,2,3,4-tetrahydro-1-naphthylamine, 1-naphthalenamine, 1,2,3,4-tetrahydro-, 1r, r-1-amino-1,2,3,4-tetrahydronaphthalene, r-1, 2, 3, 4-tetrahydro-1-naphthylamine, r-1-aminotetraline, tetralin-1alpha-amine, r-1-aminotetralin
(1R)-1,2,3,4-tetrahydronaphthalen-1-amine
Safety and Handling

Safety and Handling

TSCA : No

Documents