Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine, ChiPros™ 99+%, ee 98+%

$117.14 - $392.51

Chemical Identifiers

Chemical Name or Material (R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine
CAS 23357-46-2
Molecular Formula C10H13N
Formula Weight 147.22
MDL Number MFCD00671629
Beilstein 4231302
Synonym (R)-1-Amino-1,2,3,4-tetrahydronaphthalene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1702106

  SDS
alfa aesar™
L1702106
5g Each for $117.14

AAL1702114

  SDS
alfa aesar™
L1702114
25g Each for $392.51
Description & Specifications

Specifications

Formula Weight 147.22
Chemical Name or Material (R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine
Assay ≥99%
Molecular Formula C10H13N
MDL Number MFCD00671629
CAS 23357-46-2
Beilstein 4231302
Boiling Point 118° to 120°C (8mmHg)
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay Percent Notes ee ≥98%
Synonym (R)-1-Amino-1,2,3,4-tetrahydronaphthalene
Quantity 5g
Health Hazard 1 Warning

Applications
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries.

Solubility
Soluble in water (6.91g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.