Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-1-(1-Naphthyl)ethylamine, ChiPros 99+%, ee 98+%

$78.96 - $78.96

Chemical Identifiers

Chemical Name or Material (R)-(+)-1-(1-Naphthyl)ethylamine
CAS 3886-70-2
Molecular Formula C12H13N
Formula Weight 171.24
MDL Number MFCD00064114
UN Number UN2922
Beilstein 2208025
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1930003

  SDS
alfa aesar™
L1930003
1g Each for $78.96

AAL1930006

  SDS
alfa aesar™
L1930006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1930014

  SDS
alfa aesar™
L1930014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.24
Assay ≥99%
TSCA TSCA
Assay Percent Notes ee ≥98%
Molecular Formula C12H13N
MDL Number MFCD00064114
UN Number UN2922
Beilstein 2208025
Boiling Point 152° to 153°C (11mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Odor Amine-like
Optical Rotation +55° (c=2 in Ethanol)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318-H312
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Chemical Name or Material (R)-(+)-1-(1-Naphthyl)ethylamine
CAS 3886-70-2
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H312
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.

2° to 8°C