Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-1-(1-Naphthyl)ethylamine, 97%

Chemical Identifiers

Chemical Name or Material (R)-(+)-1-(1-Naphthyl)ethylamine
CAS 3886-70-2
Formula Weight 171.24
MDL Number MFCD00064114
UN Number UN2922
Beilstein 2208025
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6669606

  SDS
alfa aesar™
H6669606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6669614

  SDS
alfa aesar™
H6669614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.24
Boiling Point 152° to 153°C (11mmHg)
Chemical Name or Material (R)-(+)-1-(1-Naphthyl)ethylamine
Assay 97%
CAS 3886-70-2
Health Hazard 1 Danger
Physical Form Liquid
Density 1.060g/mL
MDL Number MFCD00064114
Beilstein 2208025
Odor Amine-like
Sensitivity Air Sensitive
Health Hazard 2 GHS H Statement
H301-H314-H318-H312
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Refractive Index 1.6230
UN Number UN2922
Color Colorless to pale yellow
Quantity 5g
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Applications
It is used in chiral synthesis in organic reactions including the synthesis of β-amino acids and the enantioselective of ketones to nitroolefins. Also used as absolute configuration of primary amines determined using chiral (2-nitrophenyl)proline amides and H NMR.

Solubility
Soluble in water (<10/l).

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H312
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.

2° to 8°C