missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-1-(1-Naphthyl)ethyl Isocyanate 90.0+%, TCI America™

$158.00 - $533.27

Chemical Identifiers

CAS 42340-98-7
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.237
MDL Number MFCD00004015
InChI Key GONOHGQPZFXJOJ-SNVBAGLBSA-N
Synonym 1-1-isocyanatoethyl naphthalene #, 1-1r-1-isocyanatoethyl naphthalene, 1r-1-naphthylethanisocyanate, isocyanic acid r---1-1-naphthyl ethyl ester, naphthalene,1-1r-1-isocyanatoethyl, pubchem8100, r---1-1-naphthyl ethyl isocyanate, r-1-1-naphthyl ethyl isocyanate, r-nei
PubChem CID 170633
IUPAC Name 1-[(1R)-1-isocyanatoethyl]naphthalene
SMILES CC(C1=CC=CC2=CC=CC=C21)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I03361G View Documents Tci America
I03361G
1g Each for $158.00
 
I03365G View Documents Tci America
I03365G
5g Each for $533.27
 
Specifications

Chemical Identifiers

42340-98-7
197.237
GONOHGQPZFXJOJ-SNVBAGLBSA-N
170633
CC(C1=CC=CC2=CC=CC=C21)N=C=O
C13H11NO
MFCD00004015
1-1-isocyanatoethyl naphthalene #, 1-1r-1-isocyanatoethyl naphthalene, 1r-1-naphthylethanisocyanate, isocyanic acid r---1-1-naphthyl ethyl ester, naphthalene,1-1r-1-isocyanatoethyl, pubchem8100, r---1-1-naphthyl ethyl isocyanate, r-1-1-naphthyl ethyl isocyanate, r-nei
1-[(1R)-1-isocyanatoethyl]naphthalene

Specifications

Yellow
42340-98-7
MFCD00004015
1-1-isocyanatoethyl naphthalene #, 1-1r-1-isocyanatoethyl naphthalene, 1r-1-naphthylethanisocyanate, isocyanic acid r---1-1-naphthyl ethyl ester, naphthalene,1-1r-1-isocyanatoethyl, pubchem8100, r---1-1-naphthyl ethyl isocyanate, r-1-1-naphthyl ethyl isocyanate, r-nei
CC(C1=CC=CC2=CC=CC=C21)N=C=O
197.237
197.24
≥90.0% (GC)
1g
C13H11NO
2206
GONOHGQPZFXJOJ-SNVBAGLBSA-N
1-[(1R)-1-isocyanatoethyl]naphthalene
170633
Clear Liquid at 20°C
(R)-(-)-1-(1-Naphthyl)ethyl Isocyanate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator