Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-1-(1-Naphthyl)ethyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material (R)-(-)-1-(1-Naphthyl)ethyl isocyanate
CAS 42340-98-7
Molecular Formula C13H11NO
Formula Weight 197.24
MDL Number MFCD00004015
UN Number UN2206
Beilstein 4181805
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1228603

  SDS
alfa aesar™
L1228603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL12286MD

  SDS
alfa aesar™
L12286MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.24
Chemical Name or Material (R)-(-)-1-(1-Naphthyl)ethyl isocyanate
MDL Number MFCD00004015
Molecular Formula C13H11NO
Assay 97%
Health Hazard 2 GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
UN Number UN2206
CAS 42340-98-7
Optical Rotation −47° (c=3.5 in Toluene)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Quantity 1g
Recommended Storage 2° to 8°C
Beilstein 4181805
Flash Point 93°C (199°F)
Boiling Point 106° to 108°C (0.16mmHg)
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C