missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-1-(1-Naphthyl)ethyl isocyanate, 97%

Manufacturer:  Alfa Aesar™ L1228603

 View more versions of this product

Catalog No. AAL1228603



Specifications

Specifications

(R)-(-)-1-(1-Naphthyl)ethyl isocyanate
−47° (c=3.5 in Toluene)
1g
C13H11NO
UN2206
r---1-1-naphthyl ethyl isocyanate, isocyanic acid r---1-1-naphthyl ethyl ester, 1-1r-1-isocyanatoethyl naphthalene, naphthalene,1-1r-1-isocyanatoethyl, 1r-1-naphthylethanisocyanate, 1-1-isocyanatoethyl naphthalene #, pubchem8100, r---1-1-naphthyl ethyl isocyanate, r-nei, r-1-1-naphthyl ethyl isocyanate
CC(C1=CC=CC2=CC=CC=C21)N=C=O
197.237
197.24
93°C (199°F)
106°C to 108°C (0.16mmHg)
42340-98-7
MFCD00004015
4181805
GONOHGQPZFXJOJ-SNVBAGLBSA-N
1-[(1R)-1-isocyanatoethyl]naphthalene
170633
97%

Chemical Identifiers

42340-98-7
197.237
GONOHGQPZFXJOJ-SNVBAGLBSA-N
170633
CC(C1=CC=CC2=CC=CC=C21)N=C=O
C13H11NO
MFCD00004015
r---1-1-naphthyl ethyl isocyanate, isocyanic acid r---1-1-naphthyl ethyl ester, 1-1r-1-isocyanatoethyl naphthalene, naphthalene,1-1r-1-isocyanatoethyl, 1r-1-naphthylethanisocyanate, 1-1-isocyanatoethyl naphthalene #, pubchem8100, r---1-1-naphthyl ethyl isocyanate, r-nei, r-1-1-naphthyl ethyl isocyanate
1-[(1R)-1-isocyanatoethyl]naphthalene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents