missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-1,1'-Binaphthol-2,2'-bis(trifluoromethanesulfonate), 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 350430050

 View more versions of this product

Catalog No. AC350430050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

550.44
≥96.0%
White
100.0
(R)-1, 1'-Binaphthalene-2, 2'-diyl bis(trifluoromethanesulfonate)
97%
126613-06-7
C22H12F6O6S2
-145° to -149° (20°C, 589nm) (c=1, CHCl3)
Authentic
Powder
5g
80° to 85°C
96.0
97%
(CF3SO3C10H6-)2
(R)-(-)-1, 1'-Binaphthol-2, 2'-bis(trifluoromethanesulfonate)
MFCD00274615
Glass bottle

Chemical Identifiers

126613-06-7
100.0
C22H12F6O6S2
550.44
(R)-1, 1'-Binaphthalene-2, 2'-diyl bis(trifluoromethanesulfonate)
≥96.0%
96.0
97%
(CF3SO3C10H6-)2
MFCD00274615
(R)-(-)-1, 1'-Binaphthol-2, 2'-bis(trifluoromethanesulfonate), 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA