Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-1,1'-Bi(2-naphthyl) dimethanesulfonate, 97%

Chemical Identifiers

Chemical Name or Material (R)-(-)-1,1'-Bi(2-naphthyl) dimethanesulfonate
CAS 871231-48-0
Formula Weight 442.5
MDL Number MFCD06798065
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5558903

 
alfa aesar™
H5558903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55589MD

 
alfa aesar™
H55589MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 150° to 154°C
Formula Weight 442.5
MDL Number MFCD06798065
Assay 97%
CAS 871231-48-0
Quantity 1g
Physical Form Solid
Chemical Name or Material (R)-(-)-1,1'-Bi(2-naphthyl) dimethanesulfonate