Please login to your online account to display your discounted pricing

(R)-(+)-1,1'-Bi-2-naphthol, 99%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  224302500

 View more versions of this product

Catalog No. AC224302500

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 286.32
Quantity 250mg
CAS Max % 100.0
Chemical Name or Material (R)-(+)-1, 1'-Bi-2-naphthol
Packaging Glass bottle
Linear Formula HOC10H6C10H6OH
MDL Number MFCD00004068
Molecular Formula C20H14O2
CAS 18531-94-7
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material (R)-(+)-1, 1'-Bi-2-naphthol
CAS 18531-94-7
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C20H14O2
Linear Formula HOC10H6C10H6OH
Formula Weight 286.32
MDL Number MFCD00004068
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Toxic if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents