missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quinuclidine 96.0+%, TCI America™

Manufacturer:  Tci America Q00625G

 View more versions of this product

Catalog No. Q00625G



Specifications

Specifications

Quinuclidine
5g
C7H13N
2811
SBYHFKPVCBCYGV-UHFFFAOYSA-N
1-azabicyclo[2.2.2]octane
7527
111.19
≥96.0% (GC,T)
White-Yellow
100-76-5
MFCD00006690
1,4-ethanopiperidine
C1CN2CCC1CC2
111.188
CHEBI:38420
Crystal-Powder at 20°C

Chemical Identifiers

100-76-5
111.188
SBYHFKPVCBCYGV-UHFFFAOYSA-N
7527
1-azabicyclo[2.2.2]octane
C7H13N
MFCD00006690
1,4-ethanopiperidine
CHEBI:38420
C1CN2CCC1CC2
Safety and Handling

Safety and Handling

RTECSNumber : CL5594625

TSCA : Yes

Documents