missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinuclidine, 97+%

Manufacturer:  Alfa Aesar™ H54498MD

 View more versions of this product

Catalog No. AAH54498MD



Description

Description

Applications
Quinuclidine acts as a catalyst, a chemical building block and is used in organic synthesis. It is employed to prepare quinine and alkaloids. It finds application as a ligand, which is useful in the studies of OsO4-catalyzed dihydroxylation of olefins. It plays an important role in the formation of onium salts used testing of PAC-antagonist activity.

Solubility
Soluble in alcohol, diethyl ether, water and organic solvents.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
Specifications

Specifications

Quinuclidine
157°C to 160°C
100-76-5
MFCD00006690
103111
1,4-ethanopiperidine
SBYHFKPVCBCYGV-UHFFFAOYSA-N
1-azabicyclo[2.2.2]octane
7527
111.19
Air Sensitive
250mg
C7H13N
UN2811
14,8081
Soluble in alcohol,diethyl ether,water and organic solvents.
C1CN2CCC1CC2
111.188
CHEBI:38420
≥97%

Chemical Identifiers

100-76-5
111.188
SBYHFKPVCBCYGV-UHFFFAOYSA-N
7527
1-azabicyclo[2.2.2]octane
C7H13N
MFCD00006690
1,4-ethanopiperidine
CHEBI:38420
C1CN2CCC1CC2
Safety and Handling

Safety and Handling

GHS H Statement
H301-H310-H318-H315
Toxic if swallowed.
Fatal in contact with skin.
Causes serious eye damage.
Causes skin irritation.

P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H310-H314-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 202-887-1

RTECSNumber : CL5594625

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents