Please login to your online account to display your discounted pricing

(-)-Quinpirole hydrochloride, Tocris


Selective D2-like agonist

Quantity 10mg

Manufacturer: r & d systems  106110

 View more versions of this product

Catalog No. 106110

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 255.79
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C13H21N3·HCl
Chemical Name or Material (-)-Quinpirole hydrochloride
CAS 85798-08-9
Synonym (4aR-trans)-4,4a,5,6,7,8,8a,9-Octahydro-5-propyl-1H-pyrazolo[3,4-g]quinoline hydrochloride
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material (-)-Quinpirole hydrochloride
Assay Percent Range >98%
Molecular Formula C13H21N3·HCl
Formula Weight 255.79
Synonym (4aR-trans)-4,4a,5,6,7,8,8a,9-Octahydro-5-propyl-1H-pyrazolo[3,4-g]quinoline hydrochloride
Accessories

Desiccate at -20°C