missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quinoxaline-6-boronic acid pinacol ester, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445520050
Specifications
Quinoxaline-6-boronic acid pinacol ester | |
5g | |
96% min. (GC) | |
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, 6-quinoxaline boronic acid pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxalin-6-ylboronic acid pinacol ester, quinoxaline, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, quinoxaline-6-boronic acid pinacol ester, quinoxaline-6-boronic acid, pinacol ester | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=NC=CN=C3C=C2 | |
256.11 | |
256.11 |
130°C to 145°C (0.6mmHg) | |
1167418-13-4 | |
C14H17BN2O2 | |
ZYWICCYXTGRUNM-UHFFFAOYSA-N | |
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline | |
44118298 | |
97% |
Chemical Identifiers
1167418-13-4 | |
256.11 | |
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, 6-quinoxaline boronic acid pinacol ester, 6-tetramethyl-1,3,2-dioxaborolan-2-yl quinoxaline, quinoxalin-6-ylboronic acid pinacol ester, quinoxaline, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, quinoxaline-6-boronic acid pinacol ester, quinoxaline-6-boronic acid, pinacol ester | |
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline |
C14H17BN2O2 | |
ZYWICCYXTGRUNM-UHFFFAOYSA-N | |
44118298 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=NC=CN=C3C=C2 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN.
IF IN EYES.
GHS Signal Word: Warning