Please login to your online account to display your discounted pricing

Quinoxaline-5-carboxylic acid, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC429580050

 View more versions of this product

Catalog No. AC429580050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94.0%
Formula Weight 174.16
Physical Form Powder
Quantity 5g
Color Brown
Melting Point 166° to 173°C
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C9H6N2O2
Assay Percent Range 95%
MDL Number MFCD02854494
Chemical Name or Material Quinoxaline-5-carboxylic acid
Infrared Spectrum Authentic
CAS 6924-66-9
Packaging Glass bottle

Specifications

Chemical Name or Material Quinoxaline-5-carboxylic acid
CAS 6924-66-9
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C9H6N2O2
Formula Weight 174.16
MDL Number MFCD02854494
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents