missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quinolinium Dichromate 97.0+%, TCI America™

Manufacturer:  Tci America Q005825G

 View more versions of this product

Catalog No. Q005825G



Specifications

Specifications

Quinolinium Dichromate
166°C
56549-24-7
MFCD00059108
quinolinium dichromate, acmc-209lt3
C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
476.325
476.33
≥97.0% (T)
Undesignated
25g
C18H16Cr2N2O7
1479
PYEIMZCGCWSHSV-UHFFFAOYSA-P
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium
11465689
Crystal-Powder at 20°C

Chemical Identifiers

56549-24-7
476.325
PYEIMZCGCWSHSV-UHFFFAOYSA-P
11465689
C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
C18H16Cr2N2O7
MFCD00059108
quinolinium dichromate, acmc-209lt3
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents