missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinolinium dichromate, 97%

Manufacturer:  Alfa Aesar™ H5530006

 View more versions of this product

Catalog No. AAH5530006



Specifications

Specifications

Quinolinium dichromate
164°C to 167°C (decomposition)
56549-24-7
MFCD00059108
acmc-209lt3
C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
476.325
476.33
97%
Orange to Yellow
5g
C18H16Cr2N2O7
UN3087
PYEIMZCGCWSHSV-UHFFFAOYSA-P
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium
11465689
Powder

Chemical Identifiers

56549-24-7
476.325
PYEIMZCGCWSHSV-UHFFFAOYSA-P
11465689
C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
C18H16Cr2N2O7
MFCD00059108
acmc-209lt3
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium
Safety and Handling

Safety and Handling

GHS H Statement
H272-H350-H317
May intensify fire
oxidizer.
May cause cancer.
May cause an allergic skin reaction.

P201-P202-P210-P220-P221-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c

H272-H317-H350

DOTInformation : Transport Hazard Class: 5.1; Packaging Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents