missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinolinium dichromate, 97%

Manufacturer:  Alfa Aesar™ H5530014

 View more versions of this product

Catalog No. AAH5530014



Specifications

Specifications

Quinolinium dichromate
164°C to 167°C (decomposition)
56549-24-7
MFCD00059108
acmc-209lt3
C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
476.325
476.33
97%
Orange to Yellow
25g
C18H16Cr2N2O7
UN3087
PYEIMZCGCWSHSV-UHFFFAOYSA-P
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium
11465689
Powder

Chemical Identifiers

56549-24-7
476.325
PYEIMZCGCWSHSV-UHFFFAOYSA-P
11465689
C1=CC=C2C(=C1)C=CC=[NH+]2.C1=CC=C2C(=C1)C=CC=[NH+]2.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
C18H16Cr2N2O7
MFCD00059108
acmc-209lt3
oxido-(oxido(dioxo)chromio)oxy-dioxochromium;quinolin-1-ium
Safety and Handling

Safety and Handling

GHS H Statement
H272-H350-H317
May intensify fire
oxidizer.
May cause cancer.
May cause an allergic skin reaction.

GHS P Statement
P221-P210-P220-P363-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packaging Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.

Documents