missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quinoline 97.0+%, TCI America™

Manufacturer:  Tci America Q0085500G

 View more versions of this product

Catalog No. Q0085500G



Specifications

Specifications

Quinoline
Undesignated
500g
C9H7N
2656
SMWDFEZZVXVKRB-UHFFFAOYSA-N
quinoline
7047
129.16
≥97.0% (GC)
238°C
-15°C
91-22-5
MFCD00006736
chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol
C1=CC=C2C(=C1)C=CC=N2
129.162
CHEBI:17362
Clear Liquid at 20°C

Chemical Identifiers

91-22-5
129.162
SMWDFEZZVXVKRB-UHFFFAOYSA-N
7047
quinoline
C9H7N
MFCD00006736
chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol
CHEBI:17362
C1=CC=C2C(=C1)C=CC=N2
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0794

RTECSNumber : VA9275000

TSCA : Yes

Documents