Please login to your online account to display your discounted pricing

Quinoline-N-oxide Hydrate 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Quinoline-N-oxide hydrate
Name Note 98%
CAS 1613-37-2
Assay ≥97.5% (on dry substance)
Assay Percent Range 98%
Molecular Formula C9H7NO·xH2O
Formula Weight 145.16
MDL Number MFCD00149472
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC153430050

  SDS
thermo fisher scientific
153430050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC153430250

  SDS
thermo fisher scientific
153430250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (on dry substance)
Physical Form Crystalline Powder and Chunks
Formula Weight 145.16
Quantity 5g
Color Ochre to brown
RTECS Number VC2340000
EINECS Number 216-560-6
Melting Point 48° to 53°C
Packaging Glass bottle
Health Hazard 1 HYGROSCOPIC
Water 22.5% max. (K.F.)
Infrared Spectrum Authentic
Chemical Name or Material Quinoline-N-oxide hydrate
MDL Number MFCD00149472
CAS 1613-37-2
Assay Percent Range 98%
Name Note 98%
Molecular Formula C9H7NO·xH2O
Safety and Handling

HYGROSCOPIC