missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinoline, 97%

Manufacturer:  Alfa Aesar™ A1154522

 View more versions of this product

Catalog No. AAA1154522


 

Description

Description

Applications
Quinoline is used as a chemical intermediate for making medicines and other organic compounds.  It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.

Solubility
More soluble in hot than cold water. Miscible with alcohol, ether

Notes
Dissolves sulfur, As{2}O{3}, phosphorus
Specifications

Specifications

Quinoline
97%
MFCD00006736
107477
More soluble in hot than cold water; Miscible with alcohol, ether
C1=CC=C2C(=C1)C=CC=N2
129.162
CHEBI:17362
129.16
7.3
101°C (214°F)
91-22-5
C9H7N
UN2656
14,8068
SMWDFEZZVXVKRB-UHFFFAOYSA-N
quinoline
7047
237°C
-15°C
100g

Chemical Identifiers

91-22-5
129.162
SMWDFEZZVXVKRB-UHFFFAOYSA-N
CHEBI:17362
C9H7N
MFCD00006736
7047
Safety and Handling

Safety and Handling

GHS H Statement
H311-H350-H341-H302-H315-H319
Toxic in contact with skin.
May cause cancer.
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P281-P305+P351+P338-P361-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: QUINOLINE

RTECSNumber : VA9275000

TSCA : TSCA

SDS Documents