Please login to your online account to display your discounted pricing

Quinoline, 96%, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  BPAC132190010

 View more versions of this product

Catalog No. AC132190010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid
Formula Weight 129.16
Quantity 1L
Color Clear light yellow to dark brown
Density 1.0950g/mL
Fieser 01,975; 02,356; 09,403
Synonym Benzo[b]pyridine
Chemical Name or Material Quinoline
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 96%
Molecular Formula C9H7N
Refractive Index 1.6230 to 1.6270 (20°C, 589nm)
MDL Number MFCD00006736
CAS 91-22-5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Toxic in contact with skin.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Quinoline
CAS 91-22-5
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C9H7N
Formula Weight 129.16
MDL Number MFCD00006736
Synonym Benzo[b]pyridine
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Toxic in contact with skin.
Toxic if swallowed.

SDS & Safety Documents