missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quinoline-8-sulfonyl Chloride 98.0+%, TCI America™

Manufacturer:  Tci America Q00165G

 View more versions of this product

Catalog No. Q00165G



Specifications

Specifications

Quinoline-8-sulfonyl Chloride
White-Yellow
5g
C9H6ClNO2S
3261
JUYUYCIJACTHMK-UHFFFAOYSA-N
quinoline-8-sulfonyl chloride
29220
Crystal-Powder at 20°C
306°C
128°C
18704-37-5
MFCD00006808
8-quinolinesulfonyl chloride, 8-chlorosulfonyl-1-benzazine, 8-quinolylsulfonyl chloride, quinoline-8-sulfonic acid chloride, unii-5zik3tjs9m, 8-quinolinesulfonylchloride, quinoline-8-sulphonyl chloride, 5zik3tjs9m, pubchem5873, acmc-1busp
C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2
227.662
227.66
≥98.0% (T)

Chemical Identifiers

18704-37-5
227.662
JUYUYCIJACTHMK-UHFFFAOYSA-N
29220
C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2
C9H6ClNO2S
MFCD00006808
8-quinolinesulfonyl chloride, 8-chlorosulfonyl-1-benzazine, 8-quinolylsulfonyl chloride, quinoline-8-sulfonic acid chloride, unii-5zik3tjs9m, 8-quinolinesulfonylchloride, quinoline-8-sulphonyl chloride, 5zik3tjs9m, pubchem5873, acmc-1busp
quinoline-8-sulfonyl chloride
Safety and Handling

Safety and Handling

RTECSNumber : VC2830000

TSCA : No

Documents