Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinoline-8-sulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material Quinoline-8-sulfonyl chloride
CAS 18704-37-5
Molecular Formula C9H6ClNO2S
Formula Weight 227.67
MDL Number MFCD00006808
UN Number UN3261
Beilstein 156347
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0567609

 
alfa aesar™
L0567609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0567618

 
alfa aesar™
L0567618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 227.67
Chemical Name or Material Quinoline-8-sulfonyl chloride
Assay 97%
Molecular Formula C9H6ClNO2S
MDL Number MFCD00006808
CAS 18704-37-5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
UN Number UN3261
Beilstein 156347
Boiling Point 306°C
Melting Point 127° to 130°C
Quantity 10g
RTECS Number VC2830000
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.