Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinoline-8-carboxylic acid, 98+%

$209.45 - $822.69

Chemical Identifiers

Chemical Name or Material Quinoline-8-carboxylic acid
CAS 86-59-9
Molecular Formula C10H7NO2
Formula Weight 173.17
MDL Number MFCD00047619
Beilstein 19176
Merck Index 14,8070
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1964403

  SDS
alfa aesar™
L1964403
1g Each for $209.45

AAL1964406

  SDS
alfa aesar™
L1964406
5g Each for $822.69
Description & Specifications

Specifications

Formula Weight 173.17
Chemical Name or Material Quinoline-8-carboxylic acid
MDL Number MFCD00047619
Health Hazard 1 Warning
Molecular Formula C10H7NO2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Melting Point 185° to 189°C
Beilstein 19176
Quantity 1g
Merck Index 14,8070
CAS 86-59-9

Applications
8-Quinolinecarboxylic acid may be used in the synthesis of novel oxorhenium(V) complexes incorporating quinoline and isoquinoline carboxylic acid derivatives, chiral 1,2,3,4-tetrahydroquinolinyl-oxazoline compounds, used as ligands for Ru-catalyzed asymmetric transfer hydrogenation of ketones and chiral quinolinyl-oxazoline compounds, used as ligands for Cu(II) catalyzed asymmetric cyclopropanation.

Solubility
Soluble in water. (3.1 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.