Learn More
Quinoline-8-carboxaldehyde, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1964203
Description
Quinoline-8-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsQuinoline-8-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Quinoline-8-carboxaldehyde | |
92°C to 94°C | |
MFCD00805837 | |
113039 | |
8-quinolinecarboxaldehyde, 8-quinolinecarbaldehyde, quinoline-8-carboxaldehyde, 8-quinoline carbaldehyde, 8-formylquinoline, ccris 6968, timtec-bb sbb010129, 8-quinolinecarboaldehyde, acmc-209j1k, ksc224g8j | |
OVZQVGZERAFSPI-UHFFFAOYSA-N | |
quinoline-8-carbaldehyde | |
170103 | |
≥98% |
38707-70-9 | |
C10H7NO | |
1 g | |
Air Sensitive | |
Insoluble in water. | |
C1=CC2=C(C(=C1)C=O)N=CC=C2 | |
157.172 | |
157.17 |
Chemical Identifiers
38707-70-9 | |
157.172 | |
OVZQVGZERAFSPI-UHFFFAOYSA-N | |
170103 | |
C1=CC2=C(C(=C1)C=O)N=CC=C2 |
C10H7NO | |
MFCD00805837 | |
8-quinolinecarboxaldehyde, 8-quinolinecarbaldehyde, quinoline-8-carboxaldehyde, 8-quinoline carbaldehyde, 8-formylquinoline, ccris 6968, timtec-bb sbb010129, 8-quinolinecarboaldehyde, acmc-209j1k, ksc224g8j | |
quinoline-8-carbaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only