Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinoline-5-carboxaldehyde, 97+%

$213.96 - $803.11

Chemical Identifiers

Chemical Name or Material Quinoline-5-carboxaldehyde
CAS 22934-41-4
Molecular Formula C10H7NO
Formula Weight 157.17
MDL Number MFCD00805835
Beilstein 113095
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1964103

  SDS
alfa aesar™
L1964103
1g Each for $213.96

AAL1964106

  SDS
alfa aesar™
L1964106
5g Each for $803.11
Description & Specifications

Specifications

Formula Weight 157.17
Chemical Name or Material Quinoline-5-carboxaldehyde
MDL Number MFCD00805835
Assay ≥97%
Molecular Formula C10H7NO
CAS 22934-41-4
Beilstein 113095
Health Hazard 1 Warning
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 94° to 98°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
Quinoline-5-carboxaldehyde is used as a precursor in the preparation of pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.