missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinoline-5-boronic acid, 97%

$177.01 - $484.88

Chemical Identifiers

CAS 355386-94-6
Molecular Formula C9H8BNO2
Formula Weight 172.98
MDL Number MFCD03095058
Chemical Name or Material Quinoline-5-boronic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1963903 SDS Alfa Aesar™
L1963903
1g Each for $484.88
AAL19639MD SDS Alfa Aesar™
L19639MD
250mg Each for $177.01
Specifications Description & Specifications

Chemical Identifiers

355386-94-6
172.98
Quinoline-5-boronic acid
C9H8BNO2
MFCD03095058

Specifications

355386-94-6
172.98
144° to 148°C
Quinoline-5-boronic acid
C9H8BNO2
MFCD03095058
1g
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning