missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quinoline-4-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC04601DE
Specifications
Quinoline-4-carboxylic acid | |
97% | |
5g | |
VQMSRUREDGBWKT-UHFFFAOYSA-N | |
quinoline-4-carboxylic acid | |
10243 | |
173.17 |
486-74-8 | |
C10H7NO2 | |
4-quinolinecarboxylic acid, cinchoninic acid, 4-carboxyquinoline, cinchonic acid, usaf d-2, quinoline-4-carboxylicacid, 4-quinolinecarboxylicacid, cinchoninsaure, pubchem5843, chinolin-4-carbonsaeure | |
C1=CC=C2C(=C1)C(=CC=N2)C(=O)O | |
173.171 | |
CHEBI:18311 | |
97% |
Safety and Handling
GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously
Warning