Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinoline-3-carboxylic acid, 98%

$66.74 - $229.03

Chemical Identifiers

Chemical Name or Material Quinoline-3-carboxylic acid
CAS 6480-68-8
Molecular Formula C10H7NO2
Formula Weight 173.17
MDL Number MFCD00006770
Beilstein 126542
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1513303

  SDS
alfa aesar™
L1513303
1g Each for $66.74

AAL1513306

  SDS
alfa aesar™
L1513306
5g Each for $229.03
Description & Specifications

Specifications

Formula Weight 173.17
Chemical Name or Material Quinoline-3-carboxylic acid
Molecular Formula C10H7NO2
CAS 6480-68-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00006770
Assay 98%
Quantity 1g
Beilstein 126542
Melting Point ≈283°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.