missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quinoline-3-boronic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 367730050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quinoline-3-boronic acid | |
148.0°C to 155.0°C | |
Authentic | |
Glass bottle | |
MFCD02183527 | |
3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl | |
OB(O)C1=CC2=CC=CC=C2N=C1 | |
172.98 | |
172.98 | |
Powder |
191162-39-7 | |
Brown to Orange or Pink | |
97% | |
C9H8BNO2 | |
5 g | |
YGDICLRMNDWZAK-UHFFFAOYSA-N | |
quinolin-3-ylboronic acid | |
2734663 | |
97% |
Chemical Identifiers
191162-39-7 | |
172.98 | |
YGDICLRMNDWZAK-UHFFFAOYSA-N | |
2734663 | |
OB(O)C1=CC2=CC=CC=C2N=C1 |
C9H8BNO2 | |
MFCD02183527 | |
3-quinolineboronic acid, quinoline-3-boronic acid, 3-quinolinylboronic acid, 3-quinolylboronic acid, quinolin-3-yl-3-boronic acid, 3-quinoline boronic acid, 3-quinolineboronic aid, quinolin-3-yl-boranediol, quinolin-3-yl boronic acid, boronic acid, 3-quinolinyl | |
quinolin-3-ylboronic acid |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only