Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinoline-2-thiol, 97%

Chemical Identifiers

Chemical Name or Material Quinoline-2-thiol
CAS 2637-37-8
Formula Weight 161.22
MDL Number MFCD00006751
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5574906

  SDS
alfa aesar™
H5574906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5574903

  SDS
alfa aesar™
H5574903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55749MD

  SDS
alfa aesar™
H55749MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.22
Melting Point 174° to 176°C
Chemical Name or Material Quinoline-2-thiol
TSCA TSCA
Physical Form Powder
Assay 97%
MDL Number MFCD00006751
CAS 2637-37-8
Health Hazard 1 Warning
Color Yellow to orange
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number FG7360000

Applications
2-Quinolinethiol has been used to study the electrochemical behavior of 2-Quinolinethiol using a hanging mercury drop electrode.

Solubility
Soluble in water. (50 g/L at 20°C)

Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.