missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinoline-2-thiol, 97%

Manufacturer:  Alfa Aesar™ H5574906

 View more versions of this product

Catalog No. AAH5574906



Description

Description

Applications
2-Quinolinethiol has been used to study the electrochemical behavior of 2-Quinolinethiol using a hanging mercury drop electrode.

Solubility
Soluble in water. (50 g/L at 20°C)

Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
Specifications

Specifications

Quinoline-2-thiol
174°C to 176°C
2637-37-8
MFCD00006751
Soluble in water. (50g/L at 20°C)
C1=CC=C2C(=C1)C=CC(=S)N2
161.222
161.22
97%
Yellow to Orange
5g
C9H7NS
2 1h-quinolinethione
KXZSVYHFYHTNBI-UHFFFAOYSA-N
1H-quinoline-2-thione
1800393
Powder

Chemical Identifiers

2637-37-8
161.222
KXZSVYHFYHTNBI-UHFFFAOYSA-N
1800393
C1=CC=C2C(=C1)C=CC(=S)N2
C9H7NS
MFCD00006751
2 1h-quinolinethione
1H-quinoline-2-thione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 220-132-4

RTECSNumber : FG7360000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents