Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinoline-2-carboxaldehyde, 97%

$88.15 - $88.15

Chemical Identifiers

Chemical Name or Material Quinoline-2-carboxaldehyde
CAS 5470-96-2
Molecular Formula C10H7NO
Formula Weight 157.17
MDL Number MFCD00075032
Beilstein 113061
Synonym Quinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2020206

  SDS
alfa aesar™
L2020206
5g Each for $88.15
Description & Specifications

Specifications

Formula Weight 157.17
Molecular Formula C10H7NO
Quantity 5g
MDL Number MFCD00075032
CAS 5470-96-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Beilstein 113061
Synonym Quinaldehyde
Melting Point 66° to 70°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material Quinoline-2-carboxaldehyde

Applications
2-Quinolinecarboxaldehyde was used in the preparation of 3-(2-quinolyl)-1-phenyl-2-propenone via rapid, tandem aldol-Michael reactions with the lithium, sodium and potassium enolates of acetophenone; imine-type ligands; sugar-quinoline fluorescent sensor for the detection of Hg2+ in natural water.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.