Please login to your online account to display your discounted pricing

Quinolin-2-amine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB10673DA

 View more versions of this product

Catalog No. BTB10673DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 144.18
Quantity 1g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H8N2
CAS 580-22-3
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Chemical Name or Material Quinolin-2-amine
Packaging Amber Glass Bottle
MDL Number MFCD00038079
Synonym 2-Quinolinamine
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Quinolin-2-amine
CAS 580-22-3
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H8N2
Formula Weight 144.18
MDL Number MFCD00038079
Synonym 2-Quinolinamine
Accessories

Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

SDS & Safety Documents