missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quinolin-2-amine, 97%, Maybridge

$79.92 - $79.92

Chemical Identifiers

CAS 580-22-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H8N2
Formula Weight 144.18
MDL Number MFCD00038079
Synonym 2-Quinolinamine
Chemical Name or Material Quinolin-2-amine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
BTB10673DA SDS Fisher Scientific Company
BTB10673DA
Amber Glass Bottle 1g Each for $79.92
BTB10673DE SDS Fisher Scientific Company
BTB10673DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB10673CB SDS Fisher Scientific Company
BTB10673CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

580-22-3
100.0
C9H8N2
MFCD00038079
Quinolin-2-amine
96.5
97%
144.18
2-Quinolinamine
97%

Specifications

580-22-3
100.0
97%
144.18
2-Quinolinamine
Quinolin-2-amine
96.5
Amber Glass Bottle
C9H8N2
MFCD00038079
1g
97%

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning