Please login to your online account to display your discounted pricing

Quinine sulfate dihydrate, Acros Organics


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC418780250

 View more versions of this product

Catalog No. AC418780250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 782.96
Chemical Name or Material Quinine Sulfate Dihydrate
Melting Point 153°C
Quantity 25g
Molecular Formula H2SO4·2H2O
CAS 6119-70-6
Packaging Glass bottle

Specifications

Chemical Name or Material Quinine Sulfate Dihydrate
CAS 6119-70-6
Molecular Formula H2SO4·2H2O
Formula Weight 782.96