missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quinine monohydrochloride dihydrate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 163721000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Quinine monohydrochloride dihydrate | |
6119-47-7 | |
6% to 10% (1 g, 105°C) | |
99% | |
HCl·2H2O | |
100g | |
15, 8177 | |
− 258.00 | |
Solubility in water: 62.5g/L (20 c). Other solubilities: soluble in alcohol,glycerol and chloroform,slightly soluble in ether | |
COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.O.O.Cl | |
396.92 | |
396.92 |
99% | |
122°C | |
Authentic | |
Glass bottle | |
MFCD00151248 | |
07,311 | |
− 258.00 (20.00°C c=2,0.1M HCL) | |
quinine hcl dihydrate | |
MPQKYZPYCSTMEI-BKNREGQISA-N | |
(R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride | |
124080947 | |
99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
GHS Signal Word: Warning
RUO â Research Use Only