Learn More
Quinine, anhydrous, 99% (total base), may cont. up to 5% dihydroquinine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1045918
Description
Quinine is used in photochemistry as a common fluorescence standard and as a resolving agent for chiral acids. It is also useful for treating falciparum malaria, lupus, arthritis and vivax malaria. It acts as a flavor component in tonic water and bitter lemon. It is utilized as the chiral moiety for the ligands used in sharpless asymmetric dihydroxylation. Furthermore, it catalyzes the kinetic resolution of furanones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsQuinine is used in photochemistry as a common fluorescence standard and as a resolving agent for chiral acids. It is also useful for treating falciparum malaria, lupus, arthritis and vivax malaria. It acts as a flavor component in tonic water and bitter lemon. It is utilized as the chiral moiety for the ligands used in sharpless asymmetric dihydroxylation. Furthermore, it catalyzes the kinetic resolution of furanones.
Solubility
Soluble in alcohol, chloroform and diethyl ether. Slightly soluble in water and glycerol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Quinine | |
130-95-0 | |
(total base) | |
MFCD00198096 | |
91867 | |
14,8061 | |
Soluble in alcohol,chloroform and diethyl ether. Slightly soluble in water and glycerol. | |
−166° (c=1 in ethanol) | |
(5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl)-[6-(trideuteriomethoxy)quinolin-4-yl]methanol | |
129316724 | |
99% |
Anhydrous, may cont. up to 5% dihydroquinine | |
∼177°C (decomposition) | |
C20H24N2O2 | |
50 g | |
Light Sensitive | |
1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol, quinine anhydrous, +-2,3-o-isopropylidene-l-threitol | |
LOUPRKONTZGTKE-ZCXAXGJJNA-N | |
[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C | |
324.42 | |
324.42 |
Chemical Identifiers
130-95-0 | |
324.42 | |
LOUPRKONTZGTKE-ZCXAXGJJNA-N | |
129316724 |
C20H24N2O2 | |
MFCD00198096 | |
1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol, quinine anhydrous, +-2,3-o-isopropylidene-l-threitol | |
[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C |
Safety and Handling
GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P330-P333+P313-P342+P311-P363-P501c
H302-H317
EINECSNumber : 205-003-2
RTECSNumber : VA6020000
TSCA : Yes
RUO – Research Use Only