Learn More
Quinidine sulfate dihydrate, 98+%, Thermo Scientific Chemicals
A sodium channel blocker, and to a lesser extent also blocks potassium channels
Supplier: Thermo Scientific Chemicals J6042614
Description
Quinidine sulfate dihydrate, 98+% is used as a A Na+ channel blocker.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsQuinidine sulfate dihydrate, 98+% is used as a A Na+ channel blocker.
Solubility
Soluble in chloroform, water (11 mg/ml at 20°C), alcohol, and ethanol (50 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Keep away fom strong oxidizing agents.
Specifications
Quinidine sulfate dihydrate | |
212°C to 214°C (decomposition) | |
Odorless | |
MFCD00149346 | |
Light sensitive | |
bis s-6-methoxyquinolin-4-yl 1s,2s,4s,5r-5-vinylquinuclidin-2-yl methanol sulfate dihydrate | |
ZHNFLHYOFXQIOW-CMZPEELOSA-N | |
(S)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;dihydrate | |
132274356 | |
≥98% |
6591-63-5 | |
White to Yellow | |
C40H54N4O10S | |
25 g | |
14,8060 | |
Soluble in chloroform,water (11mg/ml at 20°C),alcohol,and ethanol (50mg/ml). | |
COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.O.O.OS(=O)(=O)O | |
782.95 | |
782.94 | |
Powder |
Chemical Identifiers
6591-63-5 | |
782.95 | |
ZHNFLHYOFXQIOW-CMZPEELOSA-N | |
132274356 | |
COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.O.O.OS(=O)(=O)O |
C40H54N4O10S | |
MFCD00149346 | |
bis s-6-methoxyquinolin-4-yl 1s,2s,4s,5r-5-vinylquinuclidin-2-yl methanol sulfate dihydrate | |
(S)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;dihydrate |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 200-046-3
RTECSNumber : VA5605000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only