missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quinhydrone, TCI America™

Manufacturer:  Tci America Q0008500G

 View more versions of this product

Catalog No. Q0008500G



Specifications

Specifications

Quinhydrone
172°C
106-34-3
MFCD00010310
quinhydrone, green hydroquinone, p-benzoquinhydrone, chinhydron, chinhydron czech, unii-p4a66lq3qj, hydroquinone, compd. with p-benzoquinone, p-benzoquinone, compd. with hydroquinone, 2,5-cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol 1:1, p-benzoquinone-hydroquinone compound 1:1
C1=CC(=O)C=CC1=O.C1=CC(=CC=C1O)O
218.208
CHEBI:26491
Crystal-Powder at 20°C
Undesignated
500g
C12H10O4
2811
BDJXVNRFAQSMAA-UHFFFAOYSA-N
benzene-1,4-diol;cyclohexa-2,5-diene-1,4-dione
7801
218.21

Chemical Identifiers

106-34-3
218.208
BDJXVNRFAQSMAA-UHFFFAOYSA-N
7801
benzene-1,4-diol;cyclohexa-2,5-diene-1,4-dione
C12H10O4
MFCD00010310
quinhydrone, green hydroquinone, p-benzoquinhydrone, chinhydron, chinhydron czech, unii-p4a66lq3qj, hydroquinone, compd. with p-benzoquinone, p-benzoquinone, compd. with hydroquinone, 2,5-cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol 1:1, p-benzoquinone-hydroquinone compound 1:1
CHEBI:26491
C1=CC(=O)C=CC1=O.C1=CC(=CC=C1O)O
Safety and Handling

Safety and Handling

RTECSNumber : VA4550000

TSCA : Yes

Documents