Please login to your online account to display your discounted pricing

Quinhydrone, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  418771000

 View more versions of this product

Catalog No. AC418771000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 218.21
Physical Form Crystalline Powder
Melting Point 168° to 172°C (dec.)
Quantity 100g
Color Dark green
Merck Index 15, 8174
TSCA TSCA
RTECS Number VA4550000
EINECS Number 203-387-6
Density 1.4000g/mL
Hydroquinone 48.0 to 52.0%
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. Very toxic to aquatic life. Toxic if swallowed.
Health Hazard 1 Danger
Sulfated Ash 0.2% max.
Packaging Glass bottle
Iron (Fe) 10ppm max.
Infrared Spectrum Authentic
Sulfate 200ppm max.
p_Benzoquinone 48.0 to 52.0%
MDL Number MFCD00010310
CAS 106-34-3
Assay Percent Range 98%
Chemical Name or Material Quinhydrone
Molecular Formula C12H10O4

Specifications

Chemical Name or Material Quinhydrone
CAS 106-34-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H10O4
Formula Weight 218.21
MDL Number MFCD00010310
Accessories

Danger

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. Very toxic to aquatic life. Toxic if swallowed.

SDS & Safety Documents