missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Quincorine | |
25°C | |
270°C | |
100 mg | |
GAFZBOMPQVRGKU-FIBVVXLUSA-N | |
[(2S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]methanol | |
92020979 | |
≥98.0% (GC) |
207129-35-9 | |
White-Yellow | |
C10H17NO | |
(1S,2S,5S)-2-(Hydroxymethyl)-5-vinylquinuclidine, (1S,2S,5S)-5-Vinyl-2-quinuclidinemethanol | |
C=CC1CN2CCC1CC2CO | |
167.252 | |
167.25 | |
Crystalline Lumps |
Chemical Identifiers
207129-35-9 | |
167.252 | |
(1S,2S,5S)-2-(Hydroxymethyl)-5-vinylquinuclidine, (1S,2S,5S)-5-Vinyl-2-quinuclidinemethanol | |
[(2S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]methanol |
C10H17NO | |
GAFZBOMPQVRGKU-FIBVVXLUSA-N | |
92020979 | |
C=CC1CN2CCC1CC2CO |
Safety and Handling
TSCA : No