missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quinazoline 98%, ACROS Organics™

Manufacturer:  Acros Organics 153980010

 View more versions of this product

Catalog No. AC153980010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Quinazoline
253-82-7
Glass bottle
98%
MFCD00006712
15, 8169
C1=CC=C2C(=C1)C=NC=N2
130.15
Brown to Yellow
130.15
Crystalline Powder, Crystals and/or Chunks
98%
Authentic
≥97.5%
C8H6N2
23, 175
JWVCLYRUEFBMGU-UHFFFAOYSA-N
quinazoline
9210
CHEBI:36621
45°C to 49°C
1g

Chemical Identifiers

253-82-7
130.15
JWVCLYRUEFBMGU-UHFFFAOYSA-N
CHEBI:36621
C8H6N2
MFCD00006712
9210
Safety and Handling

Safety and Handling

EINECSNumber : 205-965-3

SDS Documents