Please login to your online account to display your discounted pricing

Alfa Aesar™ Quinazoline, 98%

$177.80 - $177.80

Chemical Identifiers

Chemical Name or Material Quinazoline
CAS 253-82-7
Molecular Formula C8H6N2
Formula Weight 130.15
MDL Number MFCD00006712
Merck Index 14,8053
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2409406

  SDS
alfa aesar™
B2409406*
5g Each for $177.80

AAB2409403

  SDS
alfa aesar™
B2409403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Quinazoline
CAS 253-82-7
Assay 98%
Molecular Formula C8H6N2
Formula Weight 130.15
MDL Number MFCD00006712
Merck Index 14,8053
Boiling Point 243°C
Melting Point 45° to 48°C
Quantity 5g
Flash Point 106°C (224°F)